EDTA RNA Storage Solution, 0.1mM
MRGF-1212GM
490014-400EA 57.87 USD
490014-400
EDTA RNA Storage Solution, 0.1mM
Comes with 1000 mL autoclavable polycarbonate bottle
Recipe: 37.22 g/L. Store at room temperature. Shelf life is three years from date of manufacture.