Would you like to log in again?
96-well Clear Bottom Black & White Microplates
Supplier: Corning
  • Nonsterile
  • Flat Well Bottom
  • Lids sold separately
Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340 nm. Opaque walls prevent well-to-well crosstalk. Optically clear flat bottom permits direct microscopic viewing. Standard 96-well plates have total well volume of 360 L, recommended working volume of 75 to 200 L. Clear bottom half area microplate has well volume of 205l, working volume of 25 to 125 L. Cell culture plates are sterilized by gamma radiation and certified non-pyrogenic.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Surface Treatment Supplier No. VWR Catalog Number Unit Price Quantity
Standard Plate TC-Treated 3720 33502-004 Case of 25 Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty