Teknova TTE, 10X
Supplier: Teknova
T4445
CA100217-738
Teknova TTE, 10X
Buffers Electrophoresis Buffers Sequencing Buffers
DNA Sequencing Grade Description: 500mM Tris, 500mM TAPS, 10mM EDTA Applications: DNA Sequencing, agarose gel electrophoresis and other applicationsrequiring TBE buffer
10 x TTE is recommended for use with the PE/ABI 377. Shipping Conditions: Ambient Storage: Room temperature