MBP MicroPoint Pipette Tip, 300 µl, Thermo Scientific
Supplier: Thermo Scientific
3571
53225-706CS 1028.21 USD
53225-706
MBP MicroPoint Pipette Tip, 300 µl, Thermo Scientific
Pipette Tips
Perform low-volume sample work with Thermo Scientific ART non-filtered pipette tips with Micropoint tip design, which feature a tip profile, nearly 50% smaller than standard blunt-end tips and significantly more narrow than beveled-end tips.
Crystal clear tips allow transparent samples to be seen easily.