Would you like to log in again?
Erie Scientific 1 mm Microscope Slide
Supplier: ERIE SCIENTIFIC SALES CO INC
Slides feature consistent 45 clipped corners for ease of handling and to aid with manual blood smears. Offering exceptional cleanliness and resistance to sticking, these slides are ideal for technicians who routinely prepare large numbers of slides. Superfrost slides have an opaque coating that takes and retains most laboratory marking methods better than plain or conventional frosted slides. The Superfrost coating is impervious to most common laboratory chemicals and reagents.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description L x W Supplier No. VWR Catalog Number Unit Price Quantity
Microscope Slide, 1 Gross Sandblasted Frosted End, 1 mm 75 x 25 mm (3 x 1") 2951 490016-282 Box of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty