NZYM Top Agar, Teknova
Supplier: Teknova
N2082 N2082
100217-512EA 175.98 USD
CA100217-512 100217-512
NZYM Top Agar, Teknova
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.