Teknova NZCYM Broth
Supplier: Teknova
N2034 N2032 N2034
100219-052EA 1497.16 USD
100219-052 CA100216-980 CA100219-052
Teknova NZCYM Broth
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.