Teknova L-Broth Agar (Miller)
Supplier: Teknova
L9240 L9240 L9235 L9235
100219-986EA 97.06 USD
CA100219-986 100219-986 CA100219-558 100219-558
Teknova L-Broth Agar (Miller)
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.