LB Agar Chloramphenicol
Supplier: Teknova
L2807 L1017 L1017 L1807 L2807 L5017 L3017 L3017 L5807 L5017
100216-574PK 175.6 USD
100216-574 CA100218-556 100217-240 CA100216-574 CA100217-062 100218-216 100218-556 CA100218-216 CA100219-306 CA100217-240 100217-062 100219-306
LB Agar Chloramphenicol
Media for Microbiology Prepared Media for Microbiology
  • Plates are Shelf-Life Dated and Guaranteed for 4 Weeks From the Date of Shipment
  • Must be Stored at 4°C and in the Absence of Continuous Exposure to Light
  • With Proper Storage, Most Antibiotic Plates are Useable for Up to 2 Months
  • All Plates are Sold in Sleeves of 20 Unless Noted Otherwise
Luria-Bertani plates contain 1% tryptone, 0.5% yeast extract, 1.0% NaCl, and 1.5% agar unless otherwise indicated. LB medium is adjusted to a final pH of 7.0 unless otherwise indicated. All plates are tested with at least 4 different recombinant strains of E. coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.