Would you like to log in again?
BioMasher™ Mini Homogenizer
Supplier: BioExpress
40012-050 40013-050
BioMasher makes protein and nucleic acid extractions easier. The system includes a micro homogenizer pestle, a filter column, and a 2.0 mL collection tube. Add your sample to the filter column, manually homogenize, and spin at 15,000 g for 10-30 seconds. Works with a wide variety of tissues and is an ideal preparatory step for many downstream applications. One-time use eliminates cross-contamination concerns. BioMasher is available with an O-ring for soft samples and overflow prevention and without an O-ring for harder tissues that need extensive grinding. Autoclavable polypropylene.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
BioMasher With O-ring 80-145 µm 40013-050 490014-284 Each Retrieving Blocked
Pack of 50 Retrieving
BioMasher Without O-ring 80-145 µm 40012-050 490014-282 Each Retrieving Blocked
Pack of 50 Retrieving
Blocked This product has been blocked by your organization.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty