Would you like to log in again?
Mo Bio Laboratories PowerMax® Soil DNA Isolation
Supplier: MO BIO LABORATORIES, INC BE
Process up to 10 grams, gives you higher yield and enriches for low copy DNAs. Purifies DNA from 10 g soil, means higher yield and more diversity of DNA from sample site. Processes more soil types including difficult environmental samples, including fecal samples. PCR inhibition was not observed in any of the samples tested. Gives high-quality DNA free of PCR inhibitors including humic substances. No visible brown coloration was seen in any of the samples tested. Ideal for biodefense, soil-borne pathogens, agriculture, and environmental studies. Vortex and centrifuge required. 1-24 Samples.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Supplier No. VWR Catalog Number Unit Price Quantity
12988-10 490015-204 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty