Would you like to log in again?
GE Nylon Graph Transfer Membranes
Supplier: GVS LIFE SCIENCES
  • Southern Transfers
  • Northern Transfers
  • Dot/Slot Blots
  • Multiple Reprobing
  • Non-Radioactive Detection Systems
  • UV Crosslinking
This membrane is optimized for low, controllable backgrounds. The GE Nylon Graph Transfer Membrane is capable of binding fragments as small as 75 bp and as great as 50 kb. It is easy to block even with procedures prone to high backgrounds such as chemiluminescent and biotinylated detection systems. This outstanding membrane provides exceptional results in both radioactive and non-radioactive detection systems. GE Nylon Graph Transfer Membranes are supported nylons, very strong, naturally hydrophilic, and have an improved binding capacity of approximately 450 µg/cm².
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Sheets 20 x 20 cm 0.22 µm 1213553 490015-616 Box of 5 Retrieving Restricted
Small Business Enterprise
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty