Magna Probe Transfer Membrane, GVS Filter Technology
1213443
490015-614BX 610.5 USD
490015-614
Magna Probe Transfer Membrane, GVS Filter Technology
Charged nylon transfer membranes.
  • Naturally hydrophilic
  • Specifically designed to allow for numerous re-probing
  • Greater binding capacity of 450 mg/cm2
  • Inherently charged to allow nucleic acid even under alkaline condition
  • Excellent sensitivity retains more nucleic acid, resulting in strong signal using smaller quantity of samples
Magna Probe charged nylon reprobing transfer membranes feature a positively charged modified nylon membrane.