Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate Sieve GQA Low Melt Agarose

Supplier: GeneMate
199-25G 199-500G
  • DNA/RNA Separation, 1 Kb
  • Analytical and Preparative Electrophoresis
  • In-gel enzymatic processing, Digestion, Ligation, PCR
  • Preparative Electrophoresis

GeneMate Sieve GQA Agarose is a low melting temperature agarose with the highest resolving capacity for DNA fragments smaller than 1000 bp, especially PCR products ranging from 200 to 800 bp. This agarose is Genetic Quality Analyzed, GQA, and carries that certification. This ensures that in-gel applications can be performed in remelted agarose, avoiding difficult DNA extraction steps. GeneMate Sieve GQA Agarose is ideal for digestion by agarose enzymes, making it very easy to recover large DNA fragments suitable for cloning or enzymatic processing. It can also be used at high concentrations, forming gels with excellent clarity and a higher sieving capacity than standard melting agaroses. GeneMate Sieve GQA gels are very easy to handle, even at concentrations as low as 3%.
Specifications:
Moisture: 5%
Ash: 0.3%
Sulfate: 0.12%
Gel Strength (4%): 1000 g/cm2
Gelling temp (4.0%): 35°C
Melting temp (4.0%): 65°C
DNase/RNase Activity: None detected
DNA Binding: None detected
In-gel enzymatic processing: Passes test
Enzymatic degradation by agarose: Passes test
Genetic Quality Testing Certified



 

ORDER

Expand All / Collapse All
Description Size VWR Catalog Number Unit Price Quantity
GeneMate LE GQA Quick Dissolve Agarose 25 g 490016-260 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty