You Searched For: Measurement Learning Activities
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (470017-032)
Supplier: VWR International
Description: Five-piece equal mass density set – for specific heat experiments and more.
Catalog Number: (470039-706)
Supplier: Avantor
Description: How Much Space Does This Take Up?
Supplier: American 3B Scientific
Description: Vibration generator for exciting oscillating and waves mechanically, such as in coil springs, a rubber cord, a wire ring or a Chladni plate. In robust...
Catalog Number: (470162-772)
Supplier: American 3B Scientific
Description: Digital counter/timer for measuring duration of motion, transition times, periods, pendulum periods, frequencies, as well as for counting events or Ge...
Catalog Number: (470072-318)
Supplier: EATON OFFICE SUPPLY
Description: WARNING: CHOKING HAZARD - Children under 8 years can choke or suffocate on uninflated or broken balloons. Adult supervision required. Keep uninflated ...
Catalog Number: (470162-778)
Supplier: American 3B Scientific
Description: Sensor for triggering the digital counter in time measurements on moving objects. Suitable for opto electronic scanning of light and dark markings on ...
Supplier: HOLIDAY HOUSE, INC.
Description: Book, How Tall How Short How Faraway, (David A. Adler)
Catalog Number: (470110-708)
Supplier: SI Manufacturing
Description: Designed to illustrate that height is proportional to mass, when diameter is uniform. Set consists of 52 identified masses which are molded to stack. ...
Catalog Number: (470162-794)
Supplier: American 3B Scientific
Description: Compact function generator
Catalog Number: (470046-274)
Supplier: Learning Resources
Description: The U.S. Customary (or English) SAFE-T Weights™ have the same features as the Metric SAFE-T Weights™.
Catalog Number: (470112-470)
Supplier: SI Manufacturing
Description: A perfect cubic decimeter box that contains exactly 1 liter. Can be used to demonstrate the metric relationships among linear, volume, one and mass me...
Supplier: SI Manufacturing
Description: Base 10 decimeter cubes. Units are 1×1×1 cm and larger pieces are scored to show units.
Catalog Number: (470014-884)
Supplier: SI Manufacturing
Description: Demonstrate the Relation of Length, Area, and Volume
Catalog Number: (470046-294)
Supplier: Learning Resources
Description: Set of three measuring pitchers with handles includes cup (8 oz/250 ml), pint (16 oz/ 250 ml), and quart (32 oz/1000 ml).
Catalog Number: (470015-982)
Supplier: Skywill Far East Industries
Description: Students gain hands-on knowledge of base 10 concepts using this set of durable plastic unit cubes, rods, flats and decimeter cubes. Utilize them in te...
Catalog Number: (470046-788)
Supplier: Learning Resources
Description: This set includes a set of four Measuring Jars with Caps, set of three Measuring Pitchers, set of five Measuring Cups, and set of five Measuring Spoon...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 17
no targeter for Bottom