Would you like to log in again?
ProSwift ConA-1S Affinity Columns, Thermo Scientific
Supplier: Thermo Scientific
074148
ORDER
Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Bed Weight Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Polymethacrylate 5 x 50 mm Concanavalin A 12 to 16 mg 074148 10040-596 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Description 10040-596
pH Range 5 to 8
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty