You Searched For: Globes
Choose from a variety of formats depicting the Earth in a detailed, three-dimensional display. Inflatable, writable globes can be used for a hands-on lesson in world geography, with the inflated globe capable of being tossed around the room and information being recorded directly on the globe through use of wet-erase markers. Mounted relief globes show mountains jutting out from the ocean-level surface of the globe, and other models include demonstrations of seasons on Earth or representations of the moon and other celestial bodies.
Choose from a variety of formats depicting the Earth in a detailed, three-dimensional display. Inflatable, writable globes can be used for a hands-on lesson in world geography, with the inflated globe capable of being tossed around the room and information being recorded directly on the globe through use of wet-erase markers. Mounted relief globes show mountains jutting out from the ocean-level surface of the globe, and other models include demonstrations of seasons on Earth or representations of the moon and other celestial bodies.
26  results were found
SearchResultCount:"26"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (470112-226)
Supplier: VWR International
Description: The Write-On Globe lets you trace the trail of Lewis and Clark, chart the discovery of the Nile, or color-code ancient empires.
Catalog Number: (470136-882)
Supplier: VWR International
Description: Get an Inside Look at Our World
Catalog Number: (470339-524)
Supplier: United Scientific Supplies
Description: A fun way to explore the countries around the globe.
Catalog Number: (470311-726)
Supplier: American 3B Scientific
Description: Tabletop globe with lighting on a metal-reinforced plastic stand with double-image map and tactile 3D relief of mountain ranges.
Catalog Number: (470324-780)
Supplier: Wards
Description: Perfect for introductory-level astronomy students.
Catalog Number: (470323-890)
Supplier: G.S.C. INTERNATIONAL, INC.
Description: This whiteboard globe has a write-on, dry-erase surface that allows for students and instructors to directly mark on it.
Catalog Number: (470150-896)
Supplier: Replogle Globes
Description: Compact, Economical Globe with Great Detail
Catalog Number: (470144-976)
Supplier: REPLOGLE GLOBES PARTNERS LLC
Description: This 9" diameter student globe is light enough to pick up and hold.
Catalog Number: (470017-010)
Supplier: PLANET EARTH GIFTS
Description: An Astronaut’s View Of the World
Catalog Number: (470004-404)
Supplier: The Science Source
Description: Economical Example of Earth’s Rotation and Seasons
Catalog Number: (470000-102)
Supplier: Science First
Description: Color-Coding for Easy Identifications
Catalog Number: (470338-910)
Supplier: United Scientific Supplies
Description: Display the apparent positions of the stars visible from Earth.
Catalog Number: (470006-238)
Supplier: Replogle Globes
Description: The Atlantis Ocean Globe
Supplier: Replogle Globes
Description: All Purpose Relief Globes indicate political divisions and illustrate topography in raised relief.
Catalog Number: (470006-250)
Supplier: Replogle Globes
Description: Wonder Globe
Catalog Number: (470004-370)
Supplier: The Science Source
Description: A Captivating Way to Show the Relationship Between the Sun and the Seasons
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 26
no targeter for Bottom