ProPac™ WAX-10 Anion Exchange Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
ProPac™
063470 088771 055150 054999 063707 063464
10044-390EA 2088.52 USD
10044-390 10044-538 10044-506 10044-504 10044-392 10044-332
ProPac™ WAX-10 Anion Exchange Protein Columns, Thermo Scientific
Chromatography Columns
For exceptionally high resolution and efficiency anion-exchange separation of protein variants, use Thermo Scientific™ ProPac SAX and WAX strong and weak anion-exchange columns.
  • Based on a nonporous 10 µm core particle that provides pressure stability, these columns can resolve isoforms that differ by a single charged residue
A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.