Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil Phenyl HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
30905-153030 30905-154030 30905-259270 30905-154630 30903-153030 30905-152130 30903-054630 30905-054630 30905-254030 30905-304030 30905-254630 30905-104630 30903-014001 30905-014001 30905-013001 30903-154630
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool

Hypersil phases are well established and referenced in many HPLC methods worldwide. They are manufactured under ISO 9001-2000 quality guidelines.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).




 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 4.6 x 250 mm 5 µm 120 Å 30905-254630 10046-098 Each Retrieving
Analytical 4 x 300 mm 5 µm 120 Å 30905-304030 10046-102 Each Retrieving
Analytical 3 x 150 mm 3 µm 120 Å 30903-153030 10046-076 Each Retrieving
Analytical 4.6 x 50 mm 5 µm 120 Å 30905-054630 10046-084 Each Retrieving
Analytical 4 x 250 mm 5 µm 120 Å 30905-254030 10046-096 Each Retrieving
Analytical 4.6 x 150 mm 3 µm 120 Å 30903-154630 10046-078 Each Retrieving
Analytical 4.6 x 100 mm 5 µm 120 Å 30905-104630 10046-086 Each Retrieving
Analytical 4.6 x 150 mm 5 µm 120 Å 30905-154630 10046-094 Each Retrieving
Analytical 4.6 x 50 mm 3 µm 120 Å 30903-054630 10046-074 Each Retrieving
Guard Cartridge 4 x 10 mm 5 µm 120 Å 30905-014001 10046-082 Each Retrieving
Guard Cartridge 4 x 10 mm 3 µm 120 Å 30903-014001 10046-072 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10046-082 10046-098 10046-102 10046-076 10046-084 10046-096 10046-078 10046-086 10046-094 10046-074 10046-072
Carbon Load 5%
pH Range 2 to 8
Surface Area 170 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty