Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil MOS-2 C8 HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
30305-152130 30305-254030 30305-154630 30303-154630 30305-054630 30305-204630 30303-014001 30305-014001 30305-034630 30303-012101 30303-104630 30305-124630 30305-254630 30305-104630
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool

Hypersil phases are well established and referenced in many HPLC methods worldwide. They are manufactured under ISO 9001-2000 quality guidelines.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).




 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 4 x 250 mm 5 µm 120 Å 30305-254030 10042-548 Each Retrieving
Analytical 4.6 x 250 mm 5 µm 120 Å 30305-254630 10042-550 Each Retrieving
Analytical 4.6 x 100 mm 3 µm 120 Å 30303-104630 10042-528 Each Retrieving
Analytical 4.6 x 150 mm 5 µm 120 Å 30305-154630 10042-544 Each Retrieving
Analytical 4.6 x 150 mm 3 µm 120 Å 30303-154630 10042-530 Each Retrieving
Analytical 4.6 x 120 mm 5 µm 120 Å 30305-124630 10042-540 Each Retrieving
Guard cartridge 4 x 10 mm 3 µm 120 Å 30303-014001 10042-526 Each Retrieving
Guard cartridge 4 x 10 mm 5 µm 120 Å 30305-014001 10042-532 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10042-548 10042-550 10042-528 10042-544 10042-530 10042-526 10042-532 10042-540
Carbon Load 6.5%
pH Range 2 to 8
Surface Area 170 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty