Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accucore 150-C18 Columns, Thermo Scientific

Supplier: Thermo Scientific
Accucore™
16126-154630 16126-102130 16126-157569 16126-104630 16126-507569 16126-014005 16126-103030 16126-033030 16126-032130 16126-034630 16126-054630 16126-053030 16126-052130 16126-013005 16126-012105 16126-153030 16126-152130
The 150A pore diameter solid core particles used in Thermo Scientific Accucore 150-C18 columns are desigend specifically to provide the optimum combination of retention and resolution for peptides.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744)




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Cartridge Guard cartridge 4.6 x 10 mm 2.6 µm 150 Å 16126-014005 10039-210 Each Retrieving
Cartridge Guard cartridge 3 x 10 mm 2.6 µm 150 Å 16126-013005 10038-912 Each Retrieving
Cartridge Guard cartridge 2.1 x 10 mm 2.6 µm 150 Å 16126-012105 10038-914 Each Retrieving
Column Analytical 4.6 x 100 mm 2.6 µm 150 Å 16126-104630 10039-192 Each Retrieving
Column Analytical 2.1 x 30 mm 2.6 µm 150 Å 16126-032130 10039-208 Each Retrieving
Column Analytical 4.6 x 150 mm 2.6 µm 150 Å 16126-154630 10039-186 Each Retrieving
Column Analytical 3 x 150 mm 2.6 µm 150 Å 16126-153030 10039-188 Each Retrieving
Column Analytical 2.1 x 100 mm 2.6 µm 150 Å 16126-102130 10039-198 Each Retrieving
Column Analytical 2.1 x 50 mm 2.6 µm 150 Å 16126-052130 10039-202 Each Retrieving
Column Analytical 4.6 x 30 mm 2.6 µm 150 Å 16126-034630 10039-204 Each Retrieving
Column Analytical 2.1 x 150 mm 2.6 µm 150 Å 16126-152130 10039-190 Each Retrieving
Column Analytical 3 x 100 mm 2.6 µm 150 Å 16126-103030 10039-194 Each Retrieving
nanoViper Column Capillary 0.075 x 150 mm 2.6 µm 150 Å 16126-157569 10039-184 Each Retrieving
nanoViper Column Capillary 0.075 x 500 mm 2.6 µm 150 Å 16126-507569 10039-182 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10039-192 10039-208 10039-210 10039-186 10039-188 10038-912 10039-198 10039-202 10039-184 10039-204 10039-190 10039-182 10039-194 10038-914
Carbon Load 7%
pH Range 1 to 11
Surface Area 80 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty