Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biobasic SCX Cation Exchange HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
73205-014001 73205-012101 73205-252130 73205-150365 73205-100365 73205-254630 73205-059570 73205-012106 73205-153030 73205-151030 73205-030315 73205-053030 73205-101030 73205-103030 73205-050565 73205-254030 73205-050365 73205-050265 73205-251030 73205-013001 73205-159070 73205-104630 73205-159370 73205-154630 73205-152130 73205-154030 73205-102130 73205-052130 73205-011001 73205-054630
For the separation of proteins, peptides, and other cationic species, choose Thermo Scientific™ BioBasic SCX columns

These high performance columns have a strong cation exchange phase based on sulfonic acid chemistry. They are ideal for the analysis of proteins, peptides, and other biomolecules, particularly for the first separation in a 2D separation.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Cartridge Guard cartridge 1 x 10 mm 5 µm 300 Å 73205-011001 10043-720 Each Retrieving
Cartridge Guard cartridge 4 x 10 mm 5 µm 300 Å 73205-014001 10043-728 Each Retrieving
Cartridge Guard cartridge 2.1 x 10 mm 5 µm 300 Å 73205-012101 10043-722 Each Retrieving
Cartridge Guard cartridge 3 x 10 mm 5 µm 300 Å 73205-013001 10043-726 Each Retrieving
Column Analytical 2.1 x 100 mm 5 µm 300 Å 73205-102130 10043-754 Each Retrieving
Column Analytical 4.6 x 100 mm 5 µm 300 Å 73205-104630 10043-758 Each Retrieving
Column Analytical 3 x 150 mm 5 µm 300 Å 73205-153030 10043-766 Each Retrieving
Column Analytical 4.6 x 250 mm 5 µm 300 Å 73205-254630 10043-782 Each Retrieving
Column Analytical 2.1 x 50 mm 5 µm 300 Å 73205-052130 10043-738 Each Retrieving
Column Analytical 3 x 100 mm 5 µm 300 Å 73205-103030 10043-756 Each Retrieving
Column Analytical 2.1 x 150 mm 5 µm 300 Å 73205-152130 10043-764 Each Retrieving
Column Preparative 10 x 150 mm 5 µm 300 Å 73205-159070 10043-772 Each Retrieving
Column Analytical 3 x 50 mm 5 µm 300 Å 73205-053030 10043-740 Each Retrieving
Column Analytical 1 x 100 mm 5 µm 300 Å 73205-101030 10043-752 Each Retrieving
Column Analytical 1 x 250 mm 5 µm 300 Å 73205-251030 10043-776 Each Retrieving
Column Analytical 4.6 x 50 mm 5 µm 300 Å 73205-054630 10043-742 Each Retrieving
Column Analytical 1 x 150 mm 5 µm 300 Å 73205-151030 10043-762 Each Retrieving
Column Analytical 4.6 x 150 mm 5 µm 300 Å 73205-154630 10043-770 Each Retrieving
Column Analytical 2.1 x 250 mm 5 µm 300 Å 73205-252130 10043-778 Each Retrieving
Javelin Guard Guard column 2.1 x 10 mm 5 µm 300 Å 73205-012106 10043-724 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10043-754 10043-724 10043-758 10043-766 10043-782 10043-720 10043-728 10043-722 10043-738 10043-756 10043-764 10043-772 10043-726 10043-740 10043-752 10043-776 10043-742 10043-762 10043-770 10043-778
Carbon Load 3%
pH Range 2 to 8
Surface Area 100 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty