Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biobasic 4 HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
72305-012101 72305-014003 72305-030215 72305-100565 72305-254630 72305-030315 72305-102168 72305-154030 72305-101030 72305-154630 72305-152130 72305-021006 72305-050565 72305-252130 72305-050265 72305-050365 72305-014001 72305-204630 72305-051030 72305-159070 72305-159270 72305-011001 72305-102130 72305-019023 72305-151030 72305-104630 72305-150365 72305-150565 72305-259070 72305-013001 72305-054630 72305-154668 72305-052130
For outstanding separation of larger peptides and proteins, choose Thermo Scientific™ BioBasic 4 HPLC Columns

These high performance columns are designed for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Cartridge Guard cartridge 10 x 10 mm 5 µm 300 Å 72305-019023 10043-580 Each Retrieving
Cartridge Guard cartridge 2.1 x 10 mm 5 µm 300 Å 72305-012101 10043-572 Each Retrieving
Cartridge Guard cartridge 4 x 10 mm 5 µm 300 Å 72305-014001 10043-576 Each Retrieving
Column Analytical 1 x 50 mm 5 µm 300 Å 72305-051030 10043-594 Each Retrieving
Column Analytical 1 x 100 mm 5 µm 300 Å 72305-101030 10043-604 Each Retrieving
Column Analytical 1 x 150 mm 5 µm 300 Å 72305-151030 10043-616 Each Retrieving
Column Analytical 2.1 x 100 mm 5 µm 300 Å 72305-102130 10043-606 Each Retrieving
Column Analytical 4.6 x 150 mm 5 µm 300 Å 72305-154630 10043-622 Each Retrieving
Column Preparative 10 x 250 mm 5 µm 300 Å 72305-259070 10043-636 Each Retrieving
Column Analytical 4.6 x 250 mm 5 µm 300 Å 72305-254630 10043-634 Each Retrieving
Column Analytical 4 x 150 mm 5 µm 300 Å 72305-154030 10043-620 Each Retrieving
Column Analytical 4.6 x 50 mm 5 µm 300 Å 72305-054630 10043-598 Each Retrieving
Column Analytical 2.1 x 150 mm 5 µm 300 Å 72305-152130 10043-618 Each Retrieving
Column Analytical 2.1 x 250 mm 5 µm 300 Å 72305-252130 10043-632 Each Retrieving
Column Analytical 2.1 x 50 mm 5 µm 300 Å 72305-052130 10043-596 Each Retrieving
Column Analytical 4.6 x 100 mm 5 µm 300 Å 72305-104630 10043-610 Each Retrieving
Javelin Guard Guard column 1 x 20 mm 5 µm 300 Å 72305-021006 10043-582 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10043-580 10043-582 10043-594 10043-604 10043-616 10043-606 10043-622 10043-636 10043-634 10043-572 10043-576 10043-620 10043-598 10043-618 10043-632 10043-596 10043-610
Carbon Load 4%
pH Range 2 to 9
Surface Area 100 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty