Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biobasic 8 HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
72205-100265 72205-014001 72205-254030 72205-102168 72205-151030 72205-152130 72205-154630 72205-051030 72205-259070 72205-052130 72205-252168 72205-011001 72205-050565 72205-012101 72205-030515 72205-012106 72205-104630 72205-030215 72205-101030 72205-254630 72205-102130
For outstanding separation of a wide range of peptides, choose Thermo Scientific™ BioBasic™ 8 HPLC Columns

BioBasic reversed-phase columns provide superior chromatography for life science applications because the extra dense bonding chemistry used for these packings produces a highly stable, reproducible surface for reliable results.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Cartridge Guard cartridge 4 x 10 mm 5 µm 300 Å 72205-014001 10043-528 Each Retrieving
Cartridge Guard cartridge 1 x 10 mm 5 µm 300 Å 72205-011001 10043-522 Each Retrieving
Cartridge Guard cartridge 2.1 x 10 mm 5 µm 300 Å 72205-012101 10043-524 Each Retrieving
Column Analytical 1 x 150 mm 5 µm 300 Å 72205-151030 10043-556 Each Retrieving
Column Analytical 1 x 50 mm 5 µm 300 Å 72205-051030 10043-536 Each Retrieving
Column Analytical 2.1 x 100 mm 5 µm 300 Å 72205-102130 10043-546 Each Retrieving
Column Analytical 2.1 x 150 mm 5 µm 300 Å 72205-152130 10043-558 Each Retrieving
Column Analytical 4 x 250 mm 5 µm 300 Å 72205-254030 10043-564 Each Retrieving
Column Capillary 0.5 x 50 mm 5 µm 300 Å 72205-050565 10043-534 Each Retrieving
Column Analytical 1 x 100 mm 5 µm 300 Å 72205-101030 10043-544 Each Retrieving
Column Analytical 4.6 x 100 mm 5 µm 300 Å 72205-104630 10043-550 Each Retrieving
Column Analytical 4.6 x 250 mm 5 µm 300 Å 72205-254630 10043-566 Each Retrieving
Column Analytical 2.1 x 50 mm 5 µm 300 Å 72205-052130 10043-538 Each Retrieving
Column Analytical 4.6 x 150 mm 5 µm 300 Å 72205-154630 10043-560 Each Retrieving
Javelin Guard Analytical 2.1 x 10 mm 5 µm 300 Å 72205-012106 10043-526 Each Retrieving
PEEK Bio-Inert Column Analytical 2.1 x 250 mm 5 µm 300 Å 72205-252168 10043-562 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10043-556 10043-528 10043-536 10043-546 10043-558 10043-564 10043-522 10043-526 10043-534 10043-544 10043-562 10043-524 10043-550 10043-566 10043-538 10043-560
Carbon Load 5%
pH Range 2 to 9
Surface Area 100 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty