Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyperREZ XP Carboyhydrate Pb²⁺ HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
69108-903027 69108-307780 69108-057726
HyperREZ XP Carbohydrate columns are based on a monodisperse resin with a 4 or 8% divinylbenzene content, and provide an ideal medium for the analysis of carbohydrates and organic acids

Unlike silica based columns they are stable at low pH, allowing the use of dilute acid as a mobile phase. The columns can also be run at elevated temperatures, for faster analysis and improved resolution of some closely eluting analytes.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical Pb²⁺ Counter Ion 7.7 x 300 mm Polymer 8 µm 69108-307780 10046-248 Each Retrieving
Guard cartridge Pb²⁺ Counter Ion 3 x 900 mm Polymer 8 µm 69108-903027 10046-250 Each Retrieving
Guard column Pb²⁺ Counter Ion 7.7 x 50 mm Polymer 8 µm 69108-057726 10046-246 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10046-246 10046-250 10046-248
pH Range 1 to 11

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty