HyperSep™ Microscale Solid Phase Extraction Tips, Thermo Scientific
Supplier: Thermo Scientific
HyperSep™
60109-207 60109-204 60109-206 60109-201 60109-218 60109-216 60109-202 60109-205 60109-215 60109-214 60109-217 60109-210 60109-208 60109-209 60109-203 60109-212 60109-213 60109-211
10047-398EA 400.78 USD
10047-398 10047-392 10047-396 10047-386 10047-420 10047-416 10047-388 10047-394 10047-414 10047-412 10047-418 10047-404 10047-400 10047-402 10047-390 10047-408 10047-410 10047-406
HyperSep™ Microscale Solid Phase Extraction Tips, Thermo Scientific
Pipet Tips
HyperSep™ tips are a revolutionary, patented series of micropipet tips designed for sample preparation.
  • Feature chromatographic media attached directly to the inner surface of the tip without using polymers or glue
  • Avoids potential problems with contamination or permeability
  • Maximizes the surface area of the chromatographic media in contact with the sample
They are ideal when working with small samples and are available with a variety of materials to suit a wide range of applications.