BioGlow® UV Transilluminators, Crystal
Supplier: Crystal
ZT-21 ZT-11 ZT-31
490015-284EA 3317.09 USD
490015-284 490006-558 490006-556
BioGlow® UV Transilluminators, Crystal
Transilluminators UV Transilluminators
The economical BioGlow® UV Transilluminators provide exceptional performance in a compact size.
  • Choice of High (100%) or Low (50%) intensity
  • Durable, hinged UV protection cover
  • Specially processed UV filter reduces background influence
  • Anti-glare surface ensured uniform UV light distribution
  • 200mm×240mm filter size to facilitate observation of large gels
Easy to maneuver, saves valuable bench top space, and have an optimized anti-glare filtered glass area with uniform illumination. Each unit is uniquely designed with a two-position intensity control device offering you both HIGH (100%) intensity and LOW (50%) intensity mode settings. This is specifically designed to meet the needs of researchers who work with both DNA and RNA. Each CTI BioGlow® UV Transilluminators comes with a UV protection cover.