Microscope Camera Systems
Supplier: Jenco
57-3M
490007-754EA 1389.6 USD
490007-754
Microscope Camera Systems
Jenco offers two types of camera systems to utilize when documenting resolved microscopic images: a digital still camera and a digital video camera. Each system is fully integrated with software for image management, optical adapters to obtain the image in the correct focal plane, and cables for connection to computers.
  • Point-to-point linear measurement
  • Area and perimeter of irregular shapes
  • Radius, area, and diameter on ellipse
  • Angle measurements
  • All components are conveniently packaged under one part number
Jenco Image archives database images and allows image annotation. It can be used to measure point-to-point or multi-pointed lengths/distances and/or area with calipers in metric, English, or pixels. The system can also calculate a color area and percentage of color relative to the whole.