The new Secador Desiccator Cabinets protect valuable reagents or samples from ambient moisture.
- Protects contents requiring dry storage conditions
- Airtight, dust- and moisture-free storage
- Automatic desiccant regeneration option
- Built-in hygrometer allows quick check on RH levels
- Transparent for easy viewing of contents
- Embedded UV protection provides longer service life
- Superior chemical resistance
- Compact footprint (WxD): 13.4" x 16.3"
- Doors open to the left
- Stackable up to 3 units high
Use to dry out specimens, pipettes/pipettors, or other components. These uniquely-designed units are clear with blue end caps. Choose either a standard Secador Desiccator Cabinet or Secador Auto-Desiccator Cabinet to meet your specific humidity protection requirements. The stackable small model is perfect for maximizing storage space in a minimum footprint. Patented dehumidifying module protects anhydrous and hygroscopic materials.