GeneMate 3 L Fernbach Flasks, TriForest
Supplier: TriForest
FBC3000S
490004-352BX 244.1 USD
490004-352
GeneMate 3 L Fernbach Flasks, TriForest
Flasks Cell Culture Flasks
GeneMate Fernbach culture flasks are sterile and designed for single use, however, they can be autoclaved and reused multiple times.
  • 45" angle is ideal for all shaker and suspension culture
  • Ideal replacement for glass or other shaker/Fernbach flasks
  • Perform aerobic & anaerobic culture
  • Certified pyrogen and nuclease free
  • Flasks are available with or without baffles
Ideal for all shaker/suspension culture.