GeneMate Individual PCR Tubes with Domed or Flat-Caps
3210-06-110IS 3210-02-110IS
490003-645BX 127.64 USD
490003-645 490003-651
GeneMate Individual PCR Tubes with Domed or Flat-Caps
  • Ultra-thin wall for optimum heat transfer (0.20 mm)
  • Certified free from DNase, RNase, and detectable DNA
These 0.2 and 0.5 mL thin-wall PCR tubes have dimensional uniformity throughout the sidewall and the conical bottom. Optimum uniform contact with the thermal cycler temperature block is ensured, resulting in consistently superior well-to-well temperature transfer and rapid cycle times. The top surface of the tube is chamfered to smoothly guide the cap to a secure locking position. The "snap seal" design gives positive and absolute assurance the tubes are fully closed and will resist popping open during thermal cycling and incubation stages. Available RNase- and DNase-free, and in natural or six different colors. Every 0.2 mL PCR tube system is designed to fit the newer generation of thermal cyclers: GeneMate, Techne, Eppendorf, Hybaid, MJ Research, Perkin-Elmer, and others. 1000 tubes per pack, cases come with 10 packs of 1000.