Sterile, non-pyrogenic, non-cytotoxic pipettes.
- Sterile, individually wrapped, and non-pyrogenic
- Lot number printed on each pipette wrapper
- Negative graduations for extended volume range
- Color-coding on wrapper and pipette allow easy size identification
- Reverse graduations for ease of reading dispensed volumes
- Accuracy within ±2% at full volume
- Polystyrene
Convenient one-side-paper, one-side-plastic packaging. Outer dust-protective bags protect during transport/storage. Colorless inert polyester-fiber plugs. Negative graduations for extended volume range. Reverse graduations for ease of reading dispensed volumes. Performance tested to ensure accuracy and integrity. Uniform top-end diameter for secure retention by controllers. Robust, uniform dispensing tips resist breakage. Uniform dispensing-tip diameters for reproducible flow rates. Crisp graduations with large easily readable numerals. Color-coded for quick volume identification. Individually Wrapped. Standard tip opening.