Brayer Roller, Diversified Biotech
Supplier: Diversified Biotech
BRAY-4000 BRAY-5000
490006-684EA 39.82 USD
490006-684 10014-782
Brayer Roller, Diversified Biotech
Film Applicators Sealing Film Applicators
The Brayer 4" Soft Rubber Roller is the perfect accessory for uniformly sealing film to microplates.