Virgin Lexan polycarbonate resin that is durable, chemically inert, non-leaching, and optically superior.Fill capacity is 20% higher than stated size. They fit all the storage areas designed for square reservoir bottle.
- Specifically designed for robotic dispensers
- Re-autoclavable (flasks and caps)
- Disposable
- Reusable
- Supplied with a standard 53B size cap
- Designed and packaged for production process or individual lab use
These are ideal for steam sterilization of all biologicals, liquids, reagents, media, and buffers.PC bottles are a safe alternative to glass, PP, or PETG bottles. They are safe, strong, transparent, leak-proof, reusable, and re-autoclavable.Optimal for all microbial culture media production and preparation of terminally sterile fluids.
Certifications: Manufactured in an ISO9001 facility with medical grade