CO2 Incubators, CB Series, O2, Tightly Sealed Divided Door, BINDER
9040-0107 9040-0106 9040-0102 9040-0110 9040-0103 9040-0111
10046-838EA 11584.38 USD
10046-838 10046-836 10046-834 10046-844 10046-842 10046-840
CO2 Incubators, CB Series, O2, Tightly Sealed Divided Door, BINDER
CO2 incubators provide optimal growth conditions with minimal condensation for cell cultures
These stackable units feature the VENTAIR™ air jacketed system delivering temperature accuracy and distribution, fast recovery times and reproducible results. The patented Permadry™ system keeps chamber walls and door dry at a maximum humidity level of 95% and the double pan humidification system provides condensation protection. Overnight the automatic hot air sterilization cycle operates at 180°C. The deep-drawn inner chamber features rounded corners and an integrated shelf support system with the unit's seamless, joint-less design preventing accumulation of particulate matter.
The units feature CO2 introduced through a patented gas mixing nozzle. The hot-air sterilizable CO2 sensor provides long term stability, a drift-free IR sensor and quick response for absolute CO2 management.
The CO2 chamber features stainless steel interiors and O2 control capable of setting the O2 content in the chamber from 0.2–95% by volume; and an airtight divided tempered safety glass door.
Models feature a and a graphical LCD color screen controller with an Ethernet interface, RS-422 interface for APT-COM™ Data and a USB port for external storage media. Additional features include the independent temperature safety device class 3.1 with visual and audible temperature alarms.
The unit has a temperature range of ambient +7°C to 60°C.
Certifications: The Control System software is compliant with GLP/GMP and FDA guideline 21 CFR Part 11.
Ordering information: The incubators are available with a platform stand and rubber pads for holding one incubator 8" off the ground. The stand is available with or without castors. Vibration-free stacking frames are also available for the direct and safe stacking of two CO2 incubators. Frames are designed on castors with stop brake and wedge equipment.