AP® Biology Investigation #8: Bacterial Transformation
Supplier: Aldon
IS3708
470062-088EA 224.99 USD
470062-088
AP® Biology Investigation #8: Bacterial Transformation
Educational Materials Biology Educational Materials AP Biology Learning Activities
Students will genetically engineer bacteria through transformation with a plasmid that confers antibiotic resistance.
  • Teachers manual and student study guide copymasters are included
  • Materials provided for 8 lab groups
They will then examine the number of resistant bacteria to determine the efficiency of the transformation procedure.
Meets AP science practices 1, 3, 5, 6, and 7, and big idea 3.