Anti-SYP Mouse Monoclonal Antibody [clone: SYP02]

Supplier: Diagnostic BioSystems

MOB399-05 MOB399
95024-822EA 309.28 USD
95024-822 99990-738
Anti-SYP Mouse Monoclonal Antibody [clone: SYP02]
Antibodies
This antibody is specific to a 38 kDa protein. Synaptophysin is an glycoprotein occurring in presynaptic vesicles of neurons in the brain, spinal cord, retina, vesicles of adrenal medulla and in neuromuscular junctions. This antibody reacts with neuroendocrine neoplasms of neural as well as epithelial types.

Type:
Antigen: SYP
Clonality: Monoclonal
Clone: SYP02
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR