Anti-PSMA Mouse Monoclonal Antibody

Supplier: Diagnostic BioSystems

MOB440-05 MOB440
99990-810EA 1808.44 USD
99990-810 95024-790
Anti-PSMA Mouse Monoclonal Antibody
Antibodies
PSMA is a 750 amino acid type II membrane glycoprotein. PSMA is expressed in normal and malignant prostatic epithelium and in a subset of non-prostatic tissues. PSMA expression has been shown to correlate with the progression of prostate cancer, with highest levels expressed in hormone-refractory and metastatic disease. PSMA expresses in the neovasculature of a variety of non-prostatic solid tumors.

Type:
Antigen: PSMA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR