Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]

Supplier: Diagnostic BioSystems

PDM014 MOB083 MOB083-05
99990-840EA 280.7 USD
99990-840 95024-778 99990-270
Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]
Antibodies
PCNA is essential for cellular DNA synthesis. This antibody reacts with proliferating cells in a wide range of normal tissues. The significance of PCNA immunoreactivity in tumors in not fully established. However, in non-Hodgkin lymphoma a linear relation between Ki-67 and PCNA staining has been demostrated.

Type:
Antigen: PCNA
Clonality: Monoclonal
Clone: PC10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR