Polysulfoethyl ASP Capillary and Micro SCX HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
164599 164566
10040-682EA 2154.88 USD
10040-682 10040-704
Polysulfoethyl ASP Capillary and Micro SCX HPLC Columns, Thermo Scientific
For very complex proteomics samples, two-dimensional LC is a powerful tool.
In the first dimension a capillary or Micro SCX column is often used in off-line and on-line 2D-LC separations. These columns are selected to establish a good top down proteomics work flow in combination with the Acclaim PepMap and Acclaim PepMap RSLC columns. Pre-installed nanoViper connection tubings allow for easy installation.