Poros 10S Capillary and Micro SCX HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
164565
10040-680EA 1858.39 USD
10040-680
Poros 10S Capillary and Micro SCX HPLC Columns, Thermo Scientific
For very complex proteomics samples, two-dimensional LC is a powerful tool.
In the first dimension a capillary or Micro SCX column is often used in off-line and on-line 2D-LC separations. These columns are selected to establish a good top down proteomics work flow in combination with the Acclaim PepMap and Acclaim PepMap RSLC columns. Pre-installed nanoViper connection tubings allow for easy installation.