Would you like to log in again?
Titanium-Dioxide Nano-Trap Columns, Thermo Scientific
Supplier: Thermo Scientific
164205 164215
Successfully enrich phosphorylated peptides from complex peptide mixtures with Thermo Scientific™ Titanium-Dioxide Nano-Trap columns prior to mass spectrometry analysis.
ORDER
Expand All / Collapse All
Column Format Phase I.D. x L Packing Material Particle Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Capillary RP 0.1 x 10 mm Titanium Dioxide 5 µm Titanium Dioxide 164205 10040-642 Each Retrieving
Capillary RP 0.2 x 10 mm Titanium Dioxide 5 µm Titanium Dioxide 164215 10040-648 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty