eVol™ XR Dispensing System, Thermo Scientific
Supplier: Thermo Scientific
66002-021 66002-024 66002-020 66002-023 66002-022
10048-570EA 2449.94 USD
10048-570 10048-564 10048-562 10048-568 10048-566
eVol™ XR Dispensing System, Thermo Scientific
The Thermo Scientific™ eVol™ Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures.
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Ideal for routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals.