Anti-HSV Type I Polyclonal Antibody

Supplier: Diagnostic BioSystems

RP018 RP018-05 PDR032
95025-048EA 308.04 USD
95025-048 99991-104 99991-232
Anti-HSV Type I Polyclonal Antibody
Antibodies
This antibody reacts with HSV type I specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.

Type:
Antigen: HSV Type I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR