Carbon Dioxide Reagent Set, Digital Titrator, Hach
Supplier: Hach
2272700 2272700
89231-368EA 121.83 USD
89231-368 CA11014-684
Carbon Dioxide Reagent Set, Digital Titrator, Hach
Environmental Test Kits
For determination of Carbon Dioxide by Digital Titrator titration. Hach Method 8205. Range: 10 - 1000 mgL as CO2 Set includes: Phenolphthalein Pillows and two Sodium Hydroxide Digital Titrator Cartridges (0.3636 N and 3.3636 N). Approximately 40-100 tests.
  • Accurate Results; when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical
Ordering information: Hazardous Items; items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.