Sodium hydroxide 1.600 N, cartridge for Digital Titrator

Supplier: Hach
Danger

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

1437901 1437901
89231-292EA 62.9 USD
CA14379-01 89231-292
Sodium hydroxide 1.600 N, cartridge for Digital Titrator
Sodium hydroxide

For acidity determination by Digital Titration, 100 - 4000 mg/L CaCO3.


  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


Ordering information: Hazardous Item; items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items

Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

EPA compliant Yes
Method 8200 (acid)
Method Name Methyl Orange
Method Number 2 8202 (P)
Method Number 3 8200 (ácido)
Method Number 4 8233 (Base)
Number of tests 40 - 100
Parameter Acidity
Platform Digital Titrator
Range 1 - 4000 mg/L CaCO3
Range 2 1 - 10 meq/L
Titrant 1.600 N NaOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR