SP Bel-Art Pipette Filler, Bel-Art Products, a part of SP
Supplier: Bel-Art
F37881-0001
13197-262EA 65.66 USD
13197-262
SP Bel-Art Pipette Filler, Bel-Art Products, a part of SP
Pipettes Pipette Bulbs
Made of Vikem® vinyl, this pipette filler has greater chemical resistance than standard rubber bulbs and accommodates pipettes from 5 to 14 mm in diameter.
  • Excellent chemical resistance
  • Vinyl fabrication
  • Easy to use
  • Three pinch valves are coded for use and can be operated with one hand
The bulb will remain collapsed when pad 'A' is released. Then, squeeze pad 'S' to aspirate liquids into the pipet. Squeezing pad 'E' will dispense liquid from the pipet.
Simply pinch valve control pad 'A' with your thumb and index finger while squeezing the bulb with three fingers and palm.