HyperSep™ Lab Plates
Supplier: Thermo Scientific
HyperSep™
60110-207 60110-301 60110-206
10047-556EA 699.63 USD
10047-556 10047-536 10047-540
HyperSep™ Lab Plates
Microplates Solid Phase Extraction Plates (SPE)
For the purification and sample preparation of proteins, DNA, RNA and other biomolecules.
  • Particle size: 20 to 40 µm
  • Pore size: 300 Å