Model 265 3 Axis Goniometer, Electron Microscopy Sciences
50176-65
100497-070EA 15583.86 USD
100497-070
Model 265 3 Axis Goniometer, Electron Microscopy Sciences
The Model 265 is a larger version of the Model 260 and is mounted on a 7" square flat base, which can be adapted for a wide range of applications.
There is ±25° of rotation in °of rotation in the other. The Model 265 is ideal for the orientation and cutting of large crystals and can easily be adapted to existing equipment. Dimensions: 7 x 7 x 5".