Model 210 Optical Orientation Instrument, Electron Microscopy Sciences
50176-10
100497-052EA 9219.42 USD
100497-052
Model 210 Optical Orientation Instrument, Electron Microscopy Sciences
The Model 210 Optical Orientation Instrument is used to determine the orientation of a crystal or to align a crystal so that specifically oriented surfaces can be cut or polished.
This uses a laser beam that is reflected off a cleaved or preferentially etched crystal surface back onto a target that is perpendicular to the laser beam. Optical orientation is an orientation technique that can be used as an alternative to more hazardous and expensive x-ray technologies.